Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002211.3 | NP_002202.2 | 3 | 350 | ||
NM_033668.2 | NP_391988.1 | 2 | 129 | |||
NM_133376.2 | NP_596867.1 | 3 | 265 | |||
GenBank mRNA | AK291697.1 | - | 3 | 202 | ||
AL710802.1 | - | 2 | 141 | |||
BC020057.1 | - | 3 | 194 | 3924411 | ||
BX537407.1 | - | 3 | 204 | |||
DB292597.1 | - | 3 | 205 |