Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014275.4 | NP_055090.1 | 9, 10 | 1421 | ||
NM_054013.3 | NP_463459.1 | 8, 9 | 1861 | |||
XM_017008998.1 | XP_016864487.1 | 9, 10 | 1247 | |||
GenBank mRNA | AB000624.1 | - | 9, 10 | 1094 | ||
AK023137.1 | - | 8, 9 | 1861 | |||
AY358984.1 | - | 9, 10 | 1102 | |||
BC009464.1 | - | 7, 8 | 748 | |||
BC051835.1 | - | 9, 10 | 1297 | 6196058 |