Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014275.4 | NP_055090.1 | 15 | 2404 | ||
NM_054013.3 | NP_463459.1 | 14 | 2844 | |||
XM_017008998.1 | XP_016864487.1 | 14 | 2326 | |||
GenBank mRNA | AK023137.1 | - | 14 | 2844 | ||
AK225490.1 | - | 12 | 1639 | |||
AK225492.1 | - | 12 | 1639 | |||
AL162067.1 | - | 12 | 1507 | |||
AY358984.1 | - | 15 | 2085 | |||
BC009464.1 | - | 13 | 1731 | |||
BC051835.1 | - | 15 | 2280 | 6196058 |