Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142466.2 | NP_001135938.1 | 5, 6 | 661 | ||
NM_133443.3 | NP_597700.1 | 5, 6 | 685 | |||
XM_017023790.1 | XP_016879279.1 | 2, 3 | 158 | |||
GenBank mRNA | AK094971.1 | - | 5, 6 | 585 | ||
AY029173.2 | - | 5, 6 | 669 | |||
BC062555.1 | - | 5, 6 | 634 | 6429257 |