Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000156.5 | NP_000147.1 | 2 | 357 | ||
NM_138924.2 | NP_620279.1 | 2 | 357 | |||
GenBank mRNA | AK289465.1 | - | 2 | 331 | ||
AK297487.1 | - | 2 | 331 | |||
BC016760.2 | - | 2 | 304 | 2989341 | ||
BC017936.1 | - | 2 | 302 | 4246521 | ||
BT007034.1 | - | 2 | 263 | |||
EU794612.1 | - | 2 | 263 | |||
Z49878.1 | - | 2 | 306 |