Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS6ST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153456.3 | NP_703157.2 | 2 | 2298 | ||
GenBank mRNA | AK126947.1 | - | 2 | 1800 | ||
CR749677.1 | - | 2 | 1963 |