Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000577.4 | NP_000568.1 | 5 | 550 | ||
NM_001318914.1 | NP_001305843.1 | 7 | 784 | |||
NM_173841.2 | NP_776213.1 | 6 | 613 | |||
NM_173842.2 | NP_776214.1 | 4 | 542 | |||
NM_173843.2 | NP_776215.1 | 6 | 721 | |||
XM_005263661.4 | XP_005263718.1 | 5 | 569 | |||
XM_011511121.1 | XP_011509423.1 | 9 | 1072 | |||
GenBank mRNA | AF043143.1 | - | 6 | 721 | ||
AK290898.1 | - | 6 | 603 | |||
AK290926.1 | - | 4 | 544 | |||
BC009745.2 | - | 5 | 455 | 3689284 | ||
BI766516.1 | - | 4 | 541 | |||
M55646.1 | - | 5 | 546 | |||
X52015.1 | - | 4 | 501 | |||
X53296.1 | - | 4 | 486 | |||
X84348.1 | - | 6 | 519 |