Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000577.4 | NP_000568.1 | 5 | 1367 | ||
NM_001318914.1 | NP_001305843.1 | 7 | 1601 | |||
NM_173841.2 | NP_776213.1 | 6 | 1430 | |||
NM_173842.2 | NP_776214.1 | 4 | 1359 | |||
NM_173843.2 | NP_776215.1 | 6 | 1538 | |||
XM_005263661.4 | XP_005263718.1 | 5 | 1386 | |||
XM_011511121.1 | XP_011509423.1 | 9 | 1889 | |||
GenBank mRNA | BC009745.2 | - | 5 | 1272 | 3689284 | |
X52015.1 | - | 4 | 1318 | |||
X53296.1 | - | 4 | 1303 |