Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193375.1 | NP_001180304.1 | 2 | 478 | ||
NM_175614.4 | NP_783313.1 | 2 | 478 | |||
NR_034166.2 | 3 | 598 | ||||
GenBank mRNA | AJ539081.1 | - | 2 | 173 | ||
AK127692.1 | - | 2 | 251 | |||
AL546175.3 | - | 3 | 339 | |||
AY726561.1 | - | 2 | 234 | |||
BC069045.1 | - | 2 | 233 | 6503302 | ||
BQ054257.1 | - | 2 | 234 | |||
BQ671956.1 | - | 2 | 252 | |||
BQ955016.1 | - | 2 | 224 |