Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178129.4 | NP_835230.1 | 2 | 1899 | ||
XM_005274429.3 | XP_005274486.1 | 2 | 2333 | |||
XM_006724443.3 | XP_006724506.2 | 3 | 2412 | |||
XM_011546178.2 | XP_011544480.1 | 3 | 2016 | |||
XM_011546179.2 | XP_011544481.1 | 2 | 1976 | |||
GenBank mRNA | AL833184.1 | - | 1 | 237 | ||
BC043610.1 | - | 2 | 1862 | 5787268 |