Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004480.4 | NP_004471.4 | 10 | 3097 | ||
NM_178155.2 | NP_835368.1 | 11 | 3527 | |||
NM_178156.2 | NP_835369.1 | 10 | 3429 | |||
NR_038167.1 | 9 | 3103 | ||||
NR_038170.1 | 11 | 2610 | ||||
XM_011536614.2 | XP_011534916.1 | 6 | 1282 | |||
XM_017021136.1 | XP_016876625.1 | 12 | 2712 | |||
XM_017021137.1 | XP_016876626.1 | 12 | 2356 | |||
XM_017021138.1 | XP_016876627.1 | 11 | 2223 | |||
XM_017021139.1 | XP_016876628.1 | 11 | 2614 | |||
XM_017021140.1 | XP_016876629.1 | 10 | 4564 | |||
GenBank mRNA | AB049740.2 | - | 6 | 1669 | ||
AK294242.1 | - | 10 | 1815 | |||
D89289.1 | - | 9 | 1942 |