Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178422.5 | NP_848509.1 | 2 | 1373 | ||
XM_005245745.3 | XP_005245802.1 | 3 | 1195 | |||
XM_005245746.3 | XP_005245803.1 | 2 | 1111 | |||
XM_011540861.1 | XP_011539163.1 | 3 | 1018 | |||
XM_011540862.2 | XP_011539164.1 | 4 | 1284 | |||
GenBank mRNA | AF313620.1 | - | 2 | 802 | ||
AY424285.1 | - | 1 | 934 | |||
BC034015.1 | - | 2 | 1373 | 4400504 | ||
BC127800.1 | - | 1 | 484 |
Set Membership: |
G-protein coupled receptor |