Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318186.1 | NP_001305115.1 | 16 | 1573 | ||
NM_001318187.1 | NP_001305116.1 | 16 | 1633 | |||
NM_001318188.1 | NP_001305117.1 | 17 | 1645 | |||
NM_002140.4 | NP_002131.2 | 17 | 1866 | |||
NM_031262.3 | NP_112552.1 | 17 | 1806 | |||
NM_031263.3 | NP_112553.1 | 17 | 1705 | |||
XM_005251960.2 | XP_005252017.1 | 17 | 1684 | |||
XM_005251963.3 | XP_005252020.1 | 16 | 1646 | |||
XM_005251965.2 | XP_005252022.1 | 16 | 1604 | |||
XM_011518616.1 | XP_011516918.1 | 16 | 1586 | |||
XM_017014668.1 | XP_016870157.1 | 17 | 1624 | |||
XM_017014669.1 | XP_016870158.1 | 16 | 1544 | |||
GenBank mRNA | AB209562.1 | - | 16 | 1562 | ||
AK096385.1 | - | 16 | 1431 | |||
AK123117.1 | - | 11 | 2371 | |||
AK294067.1 | - | 16 | 1663 | |||
AK300744.1 | - | 16 | 1516 | |||
AY911506.1 | - | 16 | 1542 | |||
BC000355.2 | - | 16 | 1601 | 2964383 | ||
BC014980.1 | - | 17 | 1612 | 4906241 | ||
BC025321.1 | - | 17 | 1564 | |||
S74678.1 | - | 16 | 1526 | |||
X72727.1 | - | 17 | 1688 |
Gene Symbol: HNRNPKP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033868.1 | 2 | 1313 | |||
GenBank mRNA | AK126814.1 | - | 2 | 1313 |