Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FFAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195755.1 | NP_001182684.1 | 1, 2 | 631 | ||
NM_181745.3 | NP_859529.2 | 1, 2 | 631 | |||
XM_011539746.2 | XP_011538048.1 | 1, 2 | 1117 | |||
GenBank mRNA | AB115768.1 | - | 1, 2 | 575 | ||
AK301525.1 | - | 1, 2 | 604 | |||
AY255573.1 | - | 1, 2 | 431 | |||
AY288417.1 | - | 1, 2 | 572 | |||
BC101175.2 | - | 1, 2 | 575 | 40016354 |
Set Membership: |
G-protein coupled receptor |