Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002143.2 | NP_002134.2 | 4 | 987 | ||
GenBank mRNA | BC001777.1 | - | 4 | 987 | 3355354 |