Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004581.5 | NP_004572.3 | 4, 5 | 931 | ||
NM_182836.2 | NP_878256.1 | 5, 6 | 614 | |||
GenBank mRNA | AK291347.1 | - | 5, 6 | 498 | ||
AK292613.1 | - | 4, 5 | 663 | |||
AK303247.1 | - | 3, 4 | 376 | |||
AK308715.1 | - | 5, 6 | 497 | |||
AL547259.3 | - | 5, 6 | 587 | |||
BC003093.1 | - | 5, 6 | 552 | 3537388 | ||
CR541797.1 | - | 3, 4 | 419 | |||
DA379718.1 | - | 5, 6 | 614 | |||
Y08200.1 | - | 4, 5 | 693 |