Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198321.3 | NP_938080.1 | 11 | 1772 | ||
GenBank mRNA | AB078145.1 | - | 11 | 1640 | ||
AJ505950.1 | - | 11 | 1640 | |||
AK023782.1 | - | 8 | 1106 | |||
AK023815.1 | - | 2 | 222 | |||
AK127135.1 | - | 11 | 1741 | |||
AL096739.1 | - | 4 | 916 | |||
BC007224.1 | - | 4 | 572 | |||
BC072450.1 | - | 7 | 1072 | |||
BC098444.1 | - | 6 | 761 |