Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198321.3 | NP_938080.1 | 12 | 4873 | ||
GenBank mRNA | AK023815.1 | - | 3 | 3323 | ||
BE875528.1 | - | 1 | 351 |