Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199141.1 | NP_954592.1 | 9, 10 | 1240 | ||
XM_005259708.4 | XP_005259765.1 | 9, 10 | 1206 | |||
XM_011527638.2 | XP_011525940.1 | 8, 9 | 996 | |||
GenBank mRNA | AF055027.1 | - | 4, 5 | 415 | ||
AL833242.1 | - | 10 | 1033 | |||
BC046240.1 | - | 537 |