Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000360.3 | NP_000351.2 | 4 | 543 | ||
NM_199292.2 | NP_954986.2 | 5 | 636 | |||
NM_199293.2 | NP_954987.2 | 5 | 624 | |||
XM_011520335.2 | XP_011518637.1 | 4 | 2831 | |||
GenBank mRNA | BC104967.1 | - | 4 | 615 | 8143970 | |
BC143611.1 | - | 4 | 613 | 9052124 | ||
BC143614.1 | - | 4 | 627 | 9052127 | ||
DQ677336.1 | - | 4 | 533 | |||
DQ677337.1 | - | 4 | 545 | |||
M17589.1 | - | 5 | 626 | |||
X05290.1 | - | 4 | 543 | |||
Y00414.1 | - | 5 | 618 |