Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000021.3 | NP_000012.1 | 4 | 430 | ||
NM_007318.2 | NP_015557.2 | 4 | 418 | |||
XM_005267864.2 | XP_005267921.1 | 4 | 424 | |||
XM_005267866.1 | XP_005267923.1 | 4 | 413 | |||
XM_011536971.2 | XP_011535273.1 | 4 | 322 | |||
XM_011536972.2 | XP_011535274.1 | 4 | 686 | |||
XM_011536973.1 | XP_011535275.1 | 4 | 674 | |||
XM_011536974.1 | XP_011535276.1 | 4 | 315 | |||
GenBank mRNA | AF416717.1 | - | 2 | 146 | ||
AJ008005.1 | - | 4 | 364 | |||
AK122722.1 | - | 2 | 1699 | |||
AK223173.1 | - | 2 | 160 | |||
AK303876.1 | - | 4 | 402 | |||
AK312531.1 | - | 4 | 414 | |||
BC011729.2 | - | 4 | 399 | 4423098 | ||
DB203403.1 | - | 4 | 418 | |||
KU178285.1 | - | 2 | 134 | |||
KU178286.1 | - | 2 | 146 | |||
L42110.1 | - | 4 | 394 | |||
L76517.1 | - | 4 | 687 | |||
U40379.1 | - | 2 | 134 | |||
U40380.1 | - | 2 | 134 |