Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000079.3 | NP_000070.1 | 5 | 523 | ||
NM_001039523.2 | NP_001034612.1 | 6 | 598 | |||
XM_017003256.1 | XP_016858745.1 | 5 | 768 | |||
XM_017003257.1 | XP_016858746.1 | 4 | 690 | |||
GenBank mRNA | AK291338.1 | - | 5 | 464 | ||
AK299445.1 | - | 6 | 587 | |||
AK315312.1 | - | 5 | 523 | |||
CD013889.1 | - | 6 | 538 | |||
S77094.1 | - | 5 | 493 | |||
Y00762.1 | - | 5 | 493 |