Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000301.3 | NP_000292.1 | 4, 5 | 523 | ||
GenBank mRNA | AK308036.1 | - | 4, 5 | 474 | ||
AK312966.1 | - | 4, 5 | 474 | |||
BC060513.1 | - | 4, 5 | 449 | 4609795 | ||
DR004070.1 | - | 4, 5 | 530 | |||
M74220.1 | - | 4, 5 | 460 | |||
X05199.1 | - | 4, 5 | 465 |
Gene Symbol: PLGLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA634686.1 | - | 4, 5 | 522 |