Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000382.2 | NP_000373.1 | 7, 8 | 1336 | ||
NM_001031806.1 | NP_001026976.1 | 7, 8 | 1336 | |||
XM_011523732.1 | XP_011522034.1 | 8, 9 | 1245 | |||
XM_011523733.1 | XP_011522035.1 | 8, 9 | 1282 | |||
XM_017024355.1 | XP_016879844.1 | 7, 8 | 1138 | |||
XM_017024356.1 | XP_016879845.1 | 8, 9 | 1244 | |||
XM_017024357.1 | XP_016879846.1 | 8, 9 | 1245 | |||
XM_017024358.1 | XP_016879847.1 | 8, 9 | 1282 | |||
GenBank mRNA | AB208894.1 | - | 7, 8 | 1044 | ||
AK000858.1 | - | 6, 7 | 946 | |||
AK025677.1 | - | 8, 9 | 1343 | |||
AK292381.1 | - | 7, 8 | 1303 | |||
AK315096.1 | - | 7, 8 | 1165 | |||
BC002430.2 | - | 7, 8 | 1156 | 3346862 | ||
CR457422.1 | - | 7, 8 | 1115 | |||
CR749559.1 | - | 4, 5 | 865 | |||
L47162.1 | - | 7, 8 | 1278 | |||
U46689.1 | - | 7, 8 | 1564 |