Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000439.4 | NP_000430.3 | 1 | 326 | ||
GenBank mRNA | BC130295.1 | - | 1 | 208 | 40146326 | |
BC136486.1 | - | 1 | 208 | 9020475 | ||
DA192592.1 | - | 1 | 326 | |||
M90753.1 | - | 1 | 296 | |||
X64810.1 | - | 1 | 276 |