Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002170.3 | NP_002161.2 | 1 | 602 | ||
GenBank mRNA | BC104828.1 | - | 1 | 601 | 8143831 | |
BC104830.1 | - | 1 | 601 | 8143833 | ||
K01900.1 | - | 1 | 616 | |||
V00545.1 | - | 1 | 603 | |||
V00550.1 | - | 1 | 603 |