Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000690.3 | NP_000681.2 | 8 | 934 | ||
NM_001204889.1 | NP_001191818.1 | 7 | 793 | |||
GenBank mRNA | AK223373.1 | - | 8 | 876 | ||
AK301375.1 | - | 7 | 731 | |||
AK314856.1 | - | 8 | 856 | |||
AY621070.1 | - | 8 | 899 | |||
BC002967.1 | - | 8 | 884 | 3543343 | ||
BC071839.1 | - | 8 | 843 | 5477768 | ||
CR456991.1 | - | 8 | 838 | |||
K03001.1 | - | 5 | 484 | |||
M26760.1 | - | 5 | 484 | |||
M54931.1 | - | 3 | 199 | |||
X05409.1 | - | 8 | 871 | |||
Y00109.1 | - | 8 | 862 |