Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000690.3 | NP_000681.2 | 13 | 1994 | ||
NM_001204889.1 | NP_001191818.1 | 12 | 1853 | |||
GenBank mRNA | AY621070.1 | - | 13 | 1959 | ||
BC071839.1 | - | 13 | 1903 | 5477768 | ||
K03001.1 | - | 10 | 1544 | |||
X05409.1 | - | 13 | 1931 | |||
Y00109.1 | - | 13 | 1922 |