Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000722.3 | NP_000713.2 | 2, 3 | 523 | ||
NM_001302890.1 | NP_001289819.1 | 2, 3 | 523 | |||
XM_005250570.2 | XP_005250627.1 | 2, 3 | 630 | |||
XM_005250572.2 | XP_005250629.1 | 2, 3 | 630 | |||
XM_005250573.2 | XP_005250630.1 | 2, 3 | 627 | |||
XM_005250574.2 | XP_005250631.1 | 2, 3 | 628 | |||
XM_006716118.2 | XP_006716181.1 | 2, 3 | 632 | |||
XM_006716119.3 | XP_006716182.1 | 2, 3 | 2415 | |||
XM_006716120.3 | XP_006716183.1 | 2, 3 | 1857 | |||
XM_011516570.2 | XP_011514872.1 | 2, 3 | 638 | |||
XM_011516571.2 | XP_011514873.1 | 2, 3 | 633 | |||
XM_011516572.2 | XP_011514874.1 | 2, 3 | 630 | |||
XM_017012588.1 | XP_016868077.1 | 2, 3 | 1856 | |||
GenBank mRNA | BC051339.1 | - | 2, 3 | 433 | ||
BC117468.1 | - | 2, 3 | 207 | 40126019 | ||
BC117470.1 | - | 2, 3 | 207 | 40126021 | ||
BP258070.1 | - | 2, 3 | 522 | |||
BU501177.1 | - | 2, 3 | 432 | |||
M76559.1 | - | 2, 3 | 210 |