Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000761.4 | NP_000752.2 | 7 | 2280 | ||
GenBank mRNA | M55053.1 | - | 6 | 2219 | ||
Z00036.1 | - | 6 | 2219 |