Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000779.3 | NP_000770.2 | 12 | 1948 | ||
NM_001099772.1 | NP_001093242.1 | 12 | 1951 | |||
NM_001319161.1 | NP_001306090.1 | 11 | 1906 | |||
NM_001319162.1 | NP_001306091.1 | 10 | 1764 | |||
NM_001319163.1 | NP_001306092.1 | 10 | 1761 | |||
NR_135003.1 | 12 | 1878 | ||||
XM_011540833.1 | XP_011539135.1 | 11 | 1584 | |||
XM_017000466.1 | XP_016855955.1 | 11 | 1855 | |||
GenBank mRNA | CD013981.1 | - | 12 | 1812 | ||
J02871.1 | - | 12 | 1877 | |||
X16699.1 | - | 12 | 1862 |