Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 4, 5 | 992 | ||
NM_001099773.1 | NP_001093243.1 | 4, 5 | 1066 | |||
GenBank mRNA | AK056794.1 | - | 4, 5 | 1061 | ||
AK292300.1 | - | 4, 5 | 898 | |||
AK292301.1 | - | 3, 4 | 856 | |||
AK293204.1 | - | 3, 4 | 698 | |||
AK293211.1 | - | 5, 6 | 858 | |||
AK300178.1 | - | 4, 5 | 808 | |||
BC032329.1 | - | 4, 5 | 881 | 5165263 | ||
KU178011.1 | - | 4, 5 | 837 | |||
M14565.1 | - | 4, 5 | 881 |