Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MME | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000902.3 | NP_000893.2 | 23 | 2575 | ||
NM_007287.2 | NP_009218.2 | 23 | 2551 | |||
NM_007288.2 | NP_009219.2 | 23 | 2597 | |||
NM_007289.2 | NP_009220.2 | 23 | 2642 | |||
XM_006713646.3 | XP_006713709.1 | 23 | 2589 | |||
XM_006713647.3 | XP_006713710.1 | 23 | 5735 | |||
XM_011512855.2 | XP_011511157.1 | 23 | 2666 | |||
XM_011512856.2 | XP_011511158.1 | 23 | 2593 | |||
XM_011512857.2 | XP_011511159.1 | 23 | 2564 | |||
GenBank mRNA | AK291761.1 | - | 23 | 2666 | ||
AL833459.1 | - | 2 | 1073 | |||
BX647134.1 | - | 23 | 2546 | |||
J03779.1 | - | 22 | 2466 | |||
X07166.1 | - | 22 | 2451 |