Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199775.1 | NP_001186704.1 | 6 | 1155 | ||
NM_001304.4 | NP_001295.2 | 6 | 1793 | |||
GenBank mRNA | AK302497.1 | - | 6 | 1397 | ||
AK302878.1 | - | 6 | 1114 | |||
AK316409.1 | - | 6 | 1155 | |||
BC045549.1 | - | 6 | 1779 | 4837034 | ||
BC045624.1 | - | 6 | 1767 | 4830195 | ||
BC051702.1 | - | 6 | 1793 | 5295685 | ||
D85390.1 | - | 6 | 1771 | |||
U65090.3 | - | 6 | 1751 |