Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001334.2 | NP_001325.1 | 2 | 248 | ||
GenBank mRNA | AK308323.1 | - | 2 | 254 | ||
AK312354.1 | - | 2 | 214 | |||
BC049206.1 | - | 2 | 201 | 5189657 | ||
BI834113.1 | - | 2 | 251 | |||
X77383.1 | - | 2 | 248 |