Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001336.3 | NP_001327.2 | 6 | 1408 | ||
GenBank mRNA | AF009923.1 | - | 6 | 1251 | ||
AF136273.1 | - | 6 | 1407 | |||
BC042168.1 | - | 6 | 1391 | 5018854 |