Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001361.4 | NP_001352.2 | 3 | 336 | ||
XM_005255827.3 | XP_005255884.1 | 3 | 641 | |||
XM_005255829.3 | XP_005255886.1 | 1 | 103 | |||
XM_017022990.1 | XP_016878479.1 | 3 | 416 | |||
GenBank mRNA | AK292293.1 | - | 4 | 492 | ||
AU130529.1 | - | 4 | 336 | |||
BC065245.1 | - | 3 | 316 | 6064723 | ||
BI253747.1 | - | 3 | 320 | |||
BM795077.1 | - | 2 | 111 | |||
M94065.1 | - | 4 | 318 |