Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301063.1 | NP_001287992.1 | 14 | 6065 | ||
NM_001543.4 | NP_001534.1 | 15 | 6149 | |||
XM_005268433.1 | XP_005268490.1 | 15 | 6160 | |||
XM_005268434.2 | XP_005268491.1 | 15 | 6769 | |||
XM_005268435.4 | XP_005268492.1 | 15 | 6294 | |||
XM_005268436.3 | XP_005268493.1 | 15 | 7934 | |||
XM_005268437.4 | XP_005268494.1 | 15 | 6173 | |||
XM_005268438.1 | XP_005268495.1 | 14 | 5989 | |||
XM_005268439.1 | XP_005268496.1 | 14 | 5950 | |||
XM_005268442.3 | XP_005268499.1 | 13 | 5143 | |||
XM_006714782.1 | XP_006714845.1 | 15 | 6195 | |||
XM_006714783.1 | XP_006714846.1 | 15 | 6121 | |||
XM_011537638.2 | XP_011535940.1 | 14 | 5250 | |||
XM_017009427.1 | XP_016864916.1 | 15 | 9151 | |||
XM_017009428.1 | XP_016864917.1 | 15 | 6254 | |||
XM_017009429.1 | XP_016864918.1 | 15 | 6134 | |||
XM_017009430.1 | XP_016864919.1 | 15 | 6156 | |||
XM_017009431.1 | XP_016864920.1 | 15 | 6726 | |||
XM_017009432.1 | XP_016864921.1 | 14 | 7946 | |||
XM_017009433.1 | XP_016864922.1 | 14 | 7905 | |||
GenBank mRNA | AB209107.1 | - | 16 | 4057 | ||
U18932.1 | - | 1 | 2918 |