Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001693.3 | NP_001684.2 | 6 | 690 | ||
GenBank mRNA | AB208806.1 | - | 6 | 480 | ||
AK294163.1 | - | 7 | 450 | |||
AK312372.1 | - | 6 | 497 | |||
BC003100.1 | - | 6 | 492 | 3502278 | ||
BC007309.1 | - | 6 | 447 | |||
BC030640.2 | - | 6 | 502 | 4821062 | ||
L35249.1 | - | 6 | 479 | |||
M60346.1 | - | 3 | 161 | |||
X62949.1 | - | 6 | 501 |