Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003962.2 | NP_001003962.1 | 9 | 864 | ||
NM_001302632.1 | NP_001289561.1 | 9 | 771 | |||
NM_001302633.1 | NP_001289562.1 | 8 | 556 | |||
NM_001749.3 | NP_001740.1 | 9 | 867 | |||
XM_005259295.1 | XP_005259352.1 | 9 | 667 | |||
XM_005259296.1 | XP_005259353.1 | 9 | 670 | |||
XM_011527359.1 | XP_011525661.1 | 8 | 497 | |||
GenBank mRNA | AK289606.1 | - | 9 | 806 | ||
AY007141.1 | - | 9 | 664 | |||
BC000592.2 | - | 9 | 685 | 3347146 | ||
BC007779.2 | - | 9 | 695 | 4301394 | ||
BC011903.2 | - | 9 | 685 | 4120579 | ||
BC017308.2 | - | 9 | 664 | 5089022 | ||
BC018931.2 | - | 9 | 755 | 4303926 | ||
BC021933.1 | - | 9 | 659 | 4872635 | ||
BC023643.2 | - | 9 | 657 | 5017512 | ||
BC064998.1 | - | 9 | 772 | 5785274 | ||
BF822052.1 | - | 3 | 151 | |||
BQ891655.1 | - | 6 | 328 | |||
BT009775.1 | - | 8 | 620 | |||
CN349928.1 | - | 8 | 505 | |||
X04106.1 | - | 9 | 778 |