Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001870.3 | NP_001861.2 | 3 | 234 | ||
GenBank mRNA | AK313628.1 | - | 3 | 194 | ||
DR005288.1 | - | 3 | 184 | |||
M27717.1 | - | 3 | 182 |