Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001870.3 | NP_001861.2 | 11 | 1194 | ||
GenBank mRNA | AK313628.1 | - | 11 | 1154 | ||
BC012613.1 | - | 11 | 1157 | 4104766 | ||
BX105358.1 | - | 1 | 97 | |||
M27717.1 | - | 11 | 1142 | |||
S40234.1 | - | 8 | 815 |