Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002203.3 | NP_002194.2 | 29 | 3548 | ||
NR_073103.1 | 28 | 3445 | ||||
NR_073104.1 | 28 | 3406 | ||||
NR_073105.1 | 29 | 3589 | ||||
NR_073106.1 | 29 | 3540 | ||||
NR_073107.1 | 28 | 3427 | ||||
GenBank mRNA | AK307952.1 | - | 29 | 3536 | ||
BC143505.1 | - | 28 | 3331 | |||
BC143508.1 | - | 28 | 3292 | |||
BC143509.1 | - | 29 | 3475 | |||
BC143511.1 | - | 29 | 3426 | |||
BC143512.1 | - | 28 | 3313 | |||
X17033.1 | - | 29 | 3453 |