Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144996.1 | NP_001138468.1 | 12, 13 | 1977 | ||
NM_001144997.1 | NP_001138469.1 | 12, 13 | 1502 | |||
NM_002206.2 | NP_002197.2 | 12, 13 | 1965 | |||
XM_005268839.1 | XP_005268896.1 | 13, 14 | 2230 | |||
XM_005268840.1 | XP_005268897.1 | 13, 14 | 2212 | |||
XM_005268841.1 | XP_005268898.1 | 13, 14 | 2230 | |||
XM_005268842.1 | XP_005268899.1 | 12, 13 | 2080 | |||
XM_005268844.1 | XP_005268901.1 | 14, 15 | 1855 | |||
XM_005268846.1 | XP_005268903.1 | 13, 14 | 1636 | |||
XM_005268848.1 | XP_005268905.1 | 12, 13 | 1654 | |||
XM_005268849.1 | XP_005268906.1 | 12, 13 | 1584 | |||
XM_005268850.1 | XP_005268907.1 | 10, 11 | 1473 | |||
XM_017019265.1 | XP_016874754.1 | 11, 12 | 1558 | |||
GenBank mRNA | AB210036.1 | - | 12, 13 | 1935 | ||
AF032108.1 | - | 12, 13 | 1906 | |||
AF052050.1 | - | 12, 13 | 1745 | |||
AF072132.1 | - | 12, 13 | 1745 | |||
AK022548.1 | - | 10, 11 | 1349 | |||
AK304864.1 | - | 12, 13 | 1478 | |||
AY358882.1 | - | 12, 13 | 1776 | |||
BC050280.1 | - | 12, 13 | 1961 | 5284443 |