Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289910.1 | NP_001276839.1 | 2 | 115 | ||
NM_002168.3 | NP_002159.2 | 2 | 348 | |||
GenBank mRNA | AK222916.1 | - | 2 | 262 | ||
AK294148.1 | - | 2 | 115 | |||
AK312627.1 | - | 2 | 312 | |||
AK316388.1 | - | 2 | 189 | |||
BC009244.2 | - | 2 | 250 | 2959540 | ||
BC071828.1 | - | 2 | 249 | 6090792 | ||
X69433.1 | - | 2 | 270 |