Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002232.4 | NP_002223.3 | 1 | 1717 | ||
NR_109845.1 | 1 | 138 | ||||
GenBank mRNA | AK314009.1 | - | 1 | 1627 | ||
BC035059.1 | - | 1 | 1718 | 5261639 | ||
BC064595.1 | - | 1 | 1440 | |||
HY035404.1 | - | 1 | 141 | |||
L23499.1 | - | 1 | 1507 | |||
M85217.1 | - | 1 | 1420 |