Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002236.4 | NP_002227.2 | 1 | 1782 | ||
GenBank mRNA | AF029780.1 | - | 1 | 1540 | ||
AF033382.1 | - | 1 | 1292 | |||
BC026110.2 | - | 1 | 1784 | 4797827 |