Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172416.1 | NP_001165887.1 | 3 | 1141 | ||
NM_001172417.1 | NP_001165888.1 | 3 | 1144 | |||
NM_002242.4 | NP_002233.2 | 3 | 1377 | |||
GenBank mRNA | AJ007557.1 | - | 3 | 1356 | ||
BC037290.2 | - | 3 | 1377 | 4817527 |