Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174097.2 | NP_001167568.1 | 3 | 542 | ||
NM_001315537.1 | NP_001302466.1 | 3 | 320 | |||
NM_002300.7 | NP_002291.1 | 3 | 320 | |||
XM_006719074.2 | XP_006719137.1 | 3 | 436 | |||
GenBank mRNA | AK129881.1 | - | 3 | 271 | ||
AK312328.1 | - | 3 | 304 | |||
AL516910.3 | - | 3 | 541 | |||
AY189689.1 | - | 3 | 296 | |||
BC002362.1 | - | 3 | 287 | 2961445 | ||
BC015122.1 | - | 3 | 311 | 3685985 | ||
BC071860.1 | - | 3 | 252 | 4902461 | ||
BT019764.1 | - | 2 | 209 | |||
BT019765.1 | - | 2 | 209 | |||
BU540074.1 | - | 3 | 323 | |||
CX867875.1 | - | 3 | 517 | |||
EU794633.1 | - | 3 | 263 | |||
Y00711.1 | - | 3 | 263 |