Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002214.2 | NP_002205.1 | 10 | 2148 | ||
XM_011515393.2 | XP_011513695.1 | 11 | 1951 | |||
XM_011515394.2 | XP_011513696.1 | 11 | 1549 | |||
XM_011515396.1 | XP_011513698.1 | 11 | 2220 | |||
XM_017012178.1 | XP_016867667.1 | 11 | 1845 | |||
XM_017012179.1 | XP_016867668.1 | 11 | 2304 | |||
XM_017012180.1 | XP_016867669.1 | 11 | 2597 | |||
XM_017012181.1 | XP_016867670.1 | 12 | 2416 | |||
XM_017012182.1 | XP_016867671.1 | 11 | 1531 | |||
XM_017012183.1 | XP_016867672.1 | 11 | 1488 | |||
GenBank mRNA | AK295044.1 | - | 11 | 2220 | ||
M73780.1 | - | 10 | 2123 |